Learn More
Timp-3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10858-1-AP-150UL
Description
The Timp-3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Timp-3. This antibody recognizes human, mouse antigen. The Timp-3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| Timp-3 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7078 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 247 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |