missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-3 Antibody (MM0036-7D3), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-61002H
This item is not returnable.
View return policy
Description
TIMP-3 Monoclonal specifically detects TIMP-3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TIMP-3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| TIMP3 | |
| Human recombinant TIMP-3 | |
| 0.1 mL | |
| Primary | |
| No cross-reactivity to human TIMP-1, 2 and 4 is noted | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0036-7D3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| Mouse | |
| Protein G purified | |
| Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
| 7078 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction