missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Rabbit anti-Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish, Clone: TIMP2/2488R, Novus Biologicals™
Rabbit Monoclonal Antibody
£440.00
Specifications
| Antigen | TIMP-2 |
|---|---|
| Clone | TIMP2/2488R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
TIMP-2 Monoclonal specifically detects TIMP-2 in Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| TIMP-2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 10 mM PBS with 0.05% BSA | |
| 7077 | |
| IgG | |
| Protein A or G purified |
| TIMP2/2488R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Guinea Pig | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| A synthetic peptide from the N-terminus of human TIMP-2 protein (aa56-66) (DSGNDIYGNPI) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title