missing translation for 'onlineSavingsMsg'
Learn More

TIMP-2 Antibody (TIMP2/2488R) - Azide and BSA Free, Novus Biologicals™

Product Code. 18378201 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378201 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378201 Supplier Novus Biologicals Supplier No. NBP308721

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TIMP-2 Monoclonal specifically detects TIMP-2 in Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish samples. It is validated for Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMP-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TIMP2/2488R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin : 1-2 ug/mL
Formulation 10mM PBS
Gene Alias CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2
Host Species Rabbit
Immunogen A synthetic peptide from the N-terminus of human TIMP-2 protein (aa56-66) (DSGNDIYGNPI)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 7077
Target Species Human, Mouse, Rat, Bovine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.