missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-2 Antibody (SPM356), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54554CL1
This item is not returnable.
View return policy
Description
TIMP-2 Monoclonal antibody specifically detects TIMP-2 in Bovine,Guinea Pig,Human,Mouse,Rabbit,Rat,Zebrafish samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Flow (Intracellular),CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| TIMP-2 | |
| Monoclonal | |
| CoraFluor 1 | |
| CSC-21Ktissue inhibitor of metalloproteinase 2, metalloproteinase inhibitor 2, TIMP metallopeptidase inhibitor 2, TIMP-2, Tissue inhibitor of metalloproteinases 2 | |
| A synthetic peptide from the N-terminus of human TIMP-2 protein (aa56-66) (DSGNDIYGNPI) (Uniprot: P16035) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers | |
| 7077 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| SPM356 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Guinea Pig, Human, Mouse, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction