missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-1 Rabbit anti-Human, Clone: TIMP1/1944R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | TIMP-1 |
|---|---|
| Clone | TIMP1/1944R |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 2-4 μg/mL |
| Applications | ELISA |
Description
TIMP-1 Monoclonal specifically detects TIMP-1 in Human samples. It is validated for ELISA.Specifications
| TIMP-1 | |
| 0.2 mg/mL | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix | |
| 10 mM PBS with 0.05% BSA | |
| 7076 | |
| IgG | |
| Protein A purified |
| TIMP1/1944R | |
| ELISA 2-4 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1 | |
| Recombinant human full-length TIMP-1 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title