missing translation for 'onlineSavingsMsg'
Learn More

TIMP-1 Antibody (SR1426), Novus Biologicals™

Rabbit Monoclonal Antibody

£265.00 - £358.00

Specifications

Antigen TIMP-1
Clone SR1426
Dilution Western Blot 1:100-1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18697514
View Documents
Novus Biologicals
NBP3-21717-100ul
100 μg
£358.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18647514
View Documents
Novus Biologicals
NBP3-21717-50ul
50 μg
£265.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TIMP-1 Monoclonal antibody specifically detects TIMP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

TIMP-1
Western Blot 1:100-1:500, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human
CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
A synthesized peptide derived from human TIMP-1 (Uniprot #: P01033)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1426
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
7076
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.