missing translation for 'onlineSavingsMsg'
Learn More

Timosaponin A1, MedChemExpress

Product Code. 16401758
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401758 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401758 Supplier Medchem Express Supplier No. HYN6079
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Timosaponin A1 is a coprostane type steroidal saponin isolated from Rhizoma Anemarrhenae.

Timosaponin A1 is a coprostane type steroidal saponin isolated from Rhizoma Anemarrhenae.

Specifications

Chemical Name or Material Timosaponin A1
CAS 68422-00-4
Color White
Purity Grade Notes Research
Molecular Formula C33H54O8
Quantity 5 mg
Solubility Information DMSO : 31.25 mg/mL (53.99 mM; ultrasonic and warming and heat to 60°C)
SMILES C[C@@]12[C@]3([H])[C@](O[C@]4(CC[C@H](C)CO4)[C@H]3C)([H])C[C@@]1([H])[C@@]5([H])[C@]([C@@]6([C@](C[C@@H](O[C@]7([H])O[C@@H]([C@H](O)[C@H](O)[C@H]7O)CO)CC6)([H])CC5)C)([H])CC2
Molecular Weight (g/mol) 578.78
Formula Weight 578.78
Percent Purity 95.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.