missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMMDC1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26071APC
This item is not returnable.
View return policy
Description
TIMMDC1 Polyclonal antibody specifically detects TIMMDC1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TIMMDC1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| chromosome 3 open reading frame 1, FLJ22597, M5-14 protein, Protein M5-14, transmembrane protein C3orf1 | |
| Produced in rabbits immunized with E. coli-derived Human TIMMDC1 fragment | |
| 0.1 mL | |
| Cell Biology | |
| 51300 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction