missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11479-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| TIMM9 | |
| Polyclonal | |
| Unconjugated | |
| TIMM9 | |
| TIM9, TIM9A, TIMM9, TIMM9A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171139, 26520, 30056 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9WV97, Q9WV98, Q9Y5J7 | |
| Timm9 | |
| TIMM9 Fusion Protein Ag2050 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction