missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM8B, Mouse anti-Human, Clone: 8E5, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00026521-M15.100ug
This item is not returnable.
View return policy
Description
Sequence: MAELGEADEAELQRLVAAEQQKAQFTAQVHHFMELCWDKCVEKPGNRLDSRTENCLSSCVDRFIDTTLAITSRFAQIVQKGGQSpecifications
| TIMM8B | |
| Monoclonal | |
| Unconjugated | |
| translocase of inner mitochondrial membrane 8 homolog B (yeast) | |
| DDP2/FLJ21744/MGC102866/MGC117373/TIM8B | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 8E5 | |
| In 1x PBS, pH 7.4 | |
| NM_012459.1 | |
| TIMM8B | |
| TIMM8B (NP_036591.1, 1 a.a. ∼ 83 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 26521 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction