missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM8A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11179-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mitochondrial intermembrane chaperone that participates in the import and insertion of some multi-pass transmembrane proteins into the mitochondrial inner membrane. Also required for the transfer of beta-barrel precursors from the TOM complex to the sorting and assembly machinery (SAM complex) of the outer membrane. Acts as a chaperone-like protein that protects the hydrophobic precursors from aggregation and guide them through the mitochondrial intermembrane space. The TIMM8-TIMM13 complex mediates the import of proteins such as TIMM23, SLC25A12/ARALAR1 and SLC25A13/ARALAR2, while the predomit TIMM9-TIMM10 70 kDa complex mediates the import of much more proteins. Probably necessary for normal neurologic development.Specifications
TIMM8A | |
Polyclonal | |
Unconjugated | |
TIMM8A | |
DDP, DDP1, Deafness dystonia protein 1, DFN1, MTS, TIM8, TIM8A, TIMM8A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1678, 30058, 84383 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.6 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60220, Q9WVA2 | |
TIMM8A, Timm8a1 | |
TIMM8A Fusion Protein Ag1661 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |