missing translation for 'onlineSavingsMsg'
Learn More

TIMM8A Antibody, Novus Biologicals™

Product Code. 18370648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370648 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370648 Supplier Novus Biologicals Supplier No. H00001678B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TIMM8A Polyclonal antibody specifically detects TIMM8A in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM8A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004076.1
Gene Alias DDP1deafness/dystonia peptide, DDPMGC12262, Deafness dystonia protein 1, DFN1, mitochondrial import inner membrane translocase subunit Tim8 A, MTSTIM8, TIM8A, translocase of inner mitochondrial membrane 8 (yeast) homolog A, translocase of inner mitochondrial membrane 8 homolog A (yeast), X-linked deafness dystonia protein
Host Species Mouse
Immunogen TIMM8A (NP_004076.1, 1 a.a. - 97 a.a.) full-length human protein. MDSSSSSSAAGLGAVDPQLQHFIEVETQKQRFQQLVHQMTELCWEKCMDKPGPKLDSRAEACFVNCVERFIDTSQFILNRLEQTQKSKPVFSESLSD
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1678
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.