missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIMM44 Recombinant Rabbit Monoclonal Antibody (K01_2J31)

Product Code. 30830325
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830325 50 μL
30827911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830325 Supplier Invitrogen™ Supplier No. MA565330
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: LPHGSTYQMR RPGGELPLSK SYSSGNRKGF LSGLLDNVKQ ELAKNKEMKE SIKKFRDEAR RLEESDVLQE ARRKYKTIES ETVRTSEVLR KKLGELTGTV KESLHEVSKS DLGRKIKEGV EEAAKTAKQS AESVSKGGEK LGRTAAFRAL SQGVESVKKE IDDSVLGQTG PYRRPQRLRK RTEFAGDKFK EEKVFEPNEE Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%, Pig - 88%, Cynomolgus monkey - 98%.

This gene encodes a peripheral membrane protein associated with the mitochondrial inner membrane translocase, which functions in the import of proteins across the mitochondrial inner membrane and into the mitochondrial matrix. The encoded protein mediates binding of mitochondrial heat shock protein 70 to the translocase of inner mitochondrial membrane 23 (TIM23) complex. Expression of this gene is upregulated in kidney in a mouse model of diabetes. A mutation in this gene is associated with familial oncocytic thyroid carcinoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIMM44
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_2J31
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene TIMM44
Gene Accession No. O43615
Gene Alias 0710005E20Rik; D8Ertd118e; Mimt44; Mitochondrial import inner membrane translocase subunit TIM44; Tim44; TIMM44; translocase of inner mitochondrial membrane 44; translocase of inner mitochondrial membrane 44 homolog; translocase of inner mitochondrial membrane 44 homolog (yeast); translocator of inner mitochondrial membrane 44; translocator of inner mitochondrial membrane 44; DNA segment, Chr 8, ERATO Doi 118, expressed
Gene Symbols TIMM44
Host Species Rabbit
Immunogen Recombinant protein of human TIMM44 (aa 30-229).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10469
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.