missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM44 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33281-20ul
This item is not returnable.
View return policy
Description
TIMM44 Monoclonal antibody specifically detects TIMM44 in Human,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TIMM44 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| MIMT44, mitochondrial import inner membrane translocase subunit TIM44, mitochondrial inner membrane translocase, TIM44DKFZp686H05241, translocase of inner mitochondrial membrane 44 homolog (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 10-240 of human TIMM44 (NP_006342.2).,, Sequence:, WCRCPRRCLGSGIQFLSSHNLPHGSTYQMRRPGGELPLSKSYSSGNRKGFLSGLLDNVKQELAKNKEMKESIKKFRDEARRLEESDVLQEARRKYKTIESETVRTSEVLRKKLGELTGTVKESLHEVSKSDLGRKIKEGVEEAAKTAKQSAESVSKGGEKLGRTAAFRALSQGVESVKKEIDDSVLGQTGPYRRPQRLRKRTEFAGDKFKEEKVFEPNEEALGVVLHKDSK | |
| 20 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 10469 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction