missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM23 Monoclonal Antibody (OTI2F5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527384
This item is not returnable.
View return policy
Description
TIMM23 Monoclonal Antibody for Western Blot, IHC (P)
Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
Specifications
| TIMM23 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14925, O35093, Q9WTQ8 | |
| Timm23 | |
| Full length human recombinant protein of TIMM23 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2F5 | |
| Unconjugated | |
| Timm23 | |
| mitochondrial import inner membrane translocase subunit Tim23; TIM23; TIMM23; translocase of inner mitochondrial membrane 23; translocase of inner mitochondrial membrane 23 homolog; translocase of inner mitochondrial membrane 23 homolog (yeast); translocase of the inner mitochondrial membrane | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 100287932, 53600, 54312 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction