missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM22 Recombinant Rabbit Monoclonal Antibody (24GB2355)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | TIMM22 |
|---|---|
| Clone | 24GB2355 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30274969
![]() |
Invitrogen™
MA557285 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274903
|
Invitrogen™
MA557286 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMM22 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
TIM22 is a translocase localized in mitochondrial inner membrane. It is implicated in import of nuclear-encoded mitochondrial preproteins to the mitochondria. TIM22 is the essential core of protein insertion complex (TIM22 complex) that consists of TIM22, TIM54, TIM18, and TIM12.Specifications
| TIMM22 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Timm22 | |
| mitochondrial import inner membrane translocase subunit Tim22; preprotein translocase; putative membrane protein; Testis-expressed protein 4; testis-expressed sequence 4; TEX4; TIM22; TIMM22; translocase of inner mitochondrial membrane 22; translocase of inner mitochondrial membrane 22 homolog; translocase of inner mitochondrial membrane 22 homolog (yeast) | |
| Timm22 | |
| Primary | |
| Affinity chromatography |
| 24GB2355 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9CQ85, Q9JKW1, Q9Y584 | |
| 29928, 56322, 79463 | |
| A synthesized peptide derived from human TIMM22 (aa 100-150). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title