missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM17B Recombinant Rabbit Monoclonal Antibody (7N3Z2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555575
This item is not returnable.
View return policy
Description
Immunogen Sequence: PQIGGSFAVW GGLFSTIDCG LVRLRGKEDP WNSITSGALT GAVLAARSGP LAMVGSAMMG GILLALIEGV GILLTRYTAQ QFRNAPPFLE DPSQLPPKDG T.
This gene encodes a multipass transmembrane protein that forms an integral component of the mitochondrial translocase TIM23 complex. This complex facilitates the transport of mitochondrial proteins from the cytosol across the mitochondrial inner membrane and into the mitochondrion. There is a pseudogene for this gene on chromosome 12. Alternative splicing results in multiple transcript variants.
Specifications
| TIMM17B | |
| Recombinant Monoclonal | |
| 1.57 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| O60830, Q9Z0V7 | |
| Timm17b | |
| A synthetic peptide corresponding to a sequence within amino acids 60-160 of human TIMM17B. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 7N3Z2 | |
| Unconjugated | |
| Timm17b | |
| 17kDa; DXS9822; inner mitochondrial membrane preprotein translocase; JM3; Mitochondrial import inner membrane translocase subunit Tim17-B; mTim17b; RGD1561744; Sfc3; TIM17B; TIMM17B; translocase of inner mitochondrial membrane 17 homolog B (yeast); translocase of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane 17 kDa, b; translocator of inner mitochondrial membrane 17b; translocator of inner mitochondrial membrane b | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10245, 21855, 317374 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction