missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM17B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94694-0.1ml
This item is not returnable.
View return policy
Description
TIMM17B Polyclonal antibody specifically detects TIMM17B in Mouse samples. It is validated for Western Blot
Specifications
| TIMM17B | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| inner mitochondrial membrane preprotein translocase, JM3, mitochondrial import inner membrane translocase subunit Tim17-B, TIM17BDXS9822, translocase of inner mitochondrial membrane 17 homolog B (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human TIMM17B (NP_005825.1). MEEYAREPCPWRIVDDCGGAFTMGVIGGGVFQAIKGFRNAPVGIRHRLRGSANAVRIRAPQIGGSFAVWGGLFSTIDCGLVRLRGKEDPWNSITSGALTGAVLAARSGPLAMVGSAMMGGILLALIEGVGILLTRYTAQQFRNAPPFLEDPSQLPPKDGTPAPGYPSYQQYH | |
| 0.1 mL | |
| Endocrinology | |
| 10245 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction