missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM17A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£231.00 - £443.00
Specifications
| Antigen | TIMM17A |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18407680
|
Novus Biologicals
NBP1-80681-25ul |
25ul |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18270766
|
Novus Biologicals
NBP1-80681 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMM17A Polyclonal specifically detects TIMM17A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TIMM17A | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q99595 | |
| 10440 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TRFASAQFPNGPQFAEDPSQLPSTQLPSSPFGDYRQYQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Inner membrane preprotein translocase Tim17a, mitochondrial import inner membrane translocase subunit Tim17-A, mitochondrial inner membrane translocase, TIM17Apreprotein translocase, TIM17MIMT17, TIMM17, translocase of inner mitochondrial membrane 17 homolog A (yeast) | |
| TIMM17A | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title