missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM17A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94024-0.1ml
This item is not returnable.
View return policy
Description
TIMM17A Polyclonal antibody specifically detects TIMM17A in Human, Mouse samples. It is validated for Western Blot
Specifications
| TIMM17A | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| Inner membrane preprotein translocase Tim17a, mitochondrial import inner membrane translocase subunit Tim17-A, mitochondrial inner membrane translocase, TIM17Apreprotein translocase, TIM17MIMT17, TIMM17, translocase of inner mitochondrial membrane 17 homolog A (yeast) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-171 of human TIMM17A (NP_006326.1). MEEYAREPCPWRIVDDCGGAFTMGTIGGGIFQAIKGFRNSPVGVNHRLRGSLTAIKTRAPQLGGSFAVWGGLFSMIDCSMVQVRGKEDPWNSITSGALTGAILAARNGPVAMVGSAAMGGILLALIEGAGILLTRFASAQFPNGPQFAEDPSQLPSTQLPSSPFGDYRQYQ | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 10440 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction