missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM13 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570051
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 100%.
This gene encodes a member of the evolutionarily conserved TIMM (translocase of inner mitochondrial membrane) family of proteins that function as chaperones in the import of proteins from the cytoplasm into the mitochondrial inner membrane. Proteins of this family play a role in collecting substrate proteins from the translocase of the outer mitochondrial membrane (TOM) complex and delivering them to either the sorting and assembly machinery in the outer mitochondrial membrane (SAM) complex or the TIMM22 complex in the inner mitochondrial membrane. The encoded protein and the translocase of mitochondrial inner membrane 8a protein form a 70 kDa complex in the intermembrane space.
Specifications
| TIMM13 | |
| Polyclonal | |
| Unconjugated | |
| TIMM13 | |
| D10Ertd378e; fd44h11; hypothetical protein LOC436738; mitochondrial import inner membrane translocase subunit Tim13; mitochondrial import inner membrane translocase subunit Tim13B; ppv1; TIM13; Tim13a; TIM13B; Tim9; Timm13; TIMM13A; TIMM13B; Timm9; translocase of inner mitochondrial membrane 13; translocase of inner mitochondrial membrane 13 homolog; translocase of inner mitochondrial membrane 13 homolog (yeast); translocase of inner mitochondrial membrane 9 homolog; wu:fd44h11; zgc:92895 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 30055 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P62075 | |
| TIMM13 | |
| synthetic peptide corresponding to a region of mouse TIMM13 (aa 5-54). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction