missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM10B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562578
This item is not returnable.
View return policy
Description
Immunogen sequence: QQQLRNLRDF LLVYNRMTEL CFQRCVPSLH HRALDAEEEA CLHSCAGKLI HSNHRLMAAY VQLMPALVQR RIADYEAASA VPGVAAEQPG VSPSGS Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.
FXC1, or TIMM10B, belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.
Specifications
| TIMM10B | |
| Polyclonal | |
| Unconjugated | |
| Timm10b | |
| fracture callus 1; fracture callus 1 homolog; fracture callus protein 1; fractured callus 1; fractured callus expressed transcript 1; FXC1; Mitochondrial import inner membrane translocase subunit Tim10 B; mitochondrial import inner membrane translocase subunit Tim9 B; small zinc finger-like protein; tim10b; Tim9b; TIMM10B; TIMM9B; translocase of inner mitochondrial membrane 10 homolog B (yeast); translocase of inner mitochondrial membrane 10B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26515 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y5J6 | |
| Timm10b | |
| Recombinant protein corresponding to Human TIMM10B. Recombinant protein control fragment (Product #RP-97328). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction