missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMM10B Monoclonal Antibody (1A11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524457
This item is not returnable.
View return policy
Description
Peptide Sequence: LRNLRDFLLV YNRMTELCFQ RCVPSLHHRA LDAEEEACLH SCAGKLIHSN HRLMAAYVQL MPALVQRRIA DYEAASAVPG VAAEQPGVSP SGS.
FXC1, or TIMM10B, belongs to a family of evolutionarily conserved proteins that are organized in heterooligomeric complexes in the mitochondrial intermembrane space. These proteins mediate the import and insertion of hydrophobic membrane proteins into the mitochondrial inner membrane.
Specifications
| TIMM10B | |
| Monoclonal | |
| Unconjugated | |
| Timm10b | |
| fracture callus 1; fracture callus 1 homolog; fracture callus protein 1; fractured callus 1; fractured callus expressed transcript 1; FXC1; Mitochondrial import inner membrane translocase subunit Tim10 B; mitochondrial import inner membrane translocase subunit Tim9 B; small zinc finger-like protein; tim10b; Tim9b; TIMM10B; TIMM9B; translocase of inner mitochondrial membrane 10 homolog B (yeast); translocase of inner mitochondrial membrane 10B | |
| Mouse | |
| Protein A | |
| RUO | |
| 26515 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1A11 | |
| PBS with no preservative; pH 7.4 | |
| Q9Y5J6 | |
| Timm10b | |
| FXC1 (NP_036324.1, 11 a.a. ~ 103 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction