missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMM10 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21445JF646
This item is not returnable.
View return policy
Description
TIMM10 Polyclonal antibody specifically detects TIMM10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TIMM10 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| mitochondrial import inner membrane translocase subunit Tim10, TIM10TIM10Atranslocase of inner mitochondrial membrane 10 (yeast) homolog, translocase of inner mitochondrial membrane 10 homolog (yeast) | |
| Produced in rabbits immunized with E. coli-derived Human TIMM10 fragment | |
| 0.1 mL | |
| Endocrinology, Signal Transduction | |
| 26519 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction