Learn More
Kartell Timer
Product Code. p-200083705
Click to view available options
Model:
Clip
Electronic
One Hand
Unit Size:
Each
Product Code. 18047761
Brand: Kartell 0090000
Only null left
Add to basket
Add to basket
Timer with ABS plastic case.
  • Portable
  • Pocket size
Specifications
Product Type Timer
Time Range 0 to 60 min.
Alarms Audible/Visible
Display Analog
Weight (English) 3.53 oz.
Weight (Metric) 0.10 kg
Model Clip