Learn More
Melford Timentin
Description
Timentin contains two antibiotics: ticarcillin (a penicillin) and clavulanate potassium (a beta-lactamase inhibitor).
Ticarcillin inhibits bacterial cell wall synthesis, disrupting the integrity of the bacterial cell wall and leading to bacterial cell lysis. Clavulanate potassium works by inhibiting beta-lactamases, enzymes produced by bacteria that can inactivate beta-lactam antibiotics, such as ticarcillin. This combination is effective against a broad spectrum of bacteria, including those producing beta-lactamases.
Specifications
Specifications
| Chemical Name or Material | Timentin |
| CAS | 4697-14-7, 61177-45-5 |
| Quantity | 5 g |
| Molecular Formula | C15H14N2Na2O6S2/C8H8KNO5 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.