Learn More
Melford Timentin
Descripción
Timentin contains two antibiotics: ticarcillin (a penicillin) and clavulanate potassium (a beta-lactamase inhibitor).
Ticarcillin inhibits bacterial cell wall synthesis, disrupting the integrity of the bacterial cell wall and leading to bacterial cell lysis. Clavulanate potassium works by inhibiting beta-lactamases, enzymes produced by bacteria that can inactivate beta-lactam antibiotics, such as ticarcillin. This combination is effective against a broad spectrum of bacteria, including those producing beta-lactamases.
Especificaciones
Especificaciones
| Chemical Name or Material | Timentin |
| CAS | 4697-14-7, 61177-45-5 |
| Quantity | 25 g |
| Molecular Formula | C15H14N2Na2O6S2/C8H8KNO5 |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.