missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMELESS Recombinant Rabbit Monoclonal Antibody (JE51-39)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534880
This item is not returnable.
View return policy
Description
Positive Control: Hela cell lysates, 293T cell lysates, Daudi cell lysates, human tonsil tissue, human colon carcinoma tissue, human gastric carcinoma tissue, human small intestine tissue, human pancreas tissue.
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
Specifications
| TIMELESS | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9UNS1 | |
| TIMELESS | |
| Recombinant protein within Human Timeless aa 1,018-1,208. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE51-39 | |
| Unconjugated | |
| TIMELESS | |
| C77407; Debt69; hTIM; mTim; Protein timeless homolog; RGD:620939}; rTIM; rTLP; TIM; Tim1; timeless; timeless {ECO:0000312; timeless circadian clock; timeless circadian clock 1; timeless circadian regulator; timeless homolog; TIMELESS1; timeless-like protein; Tof1 homolog; unknown | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8914 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction