missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIMELESS Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116046
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TIMELESS.
The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
Specifications
| TIMELESS | |
| Polyclonal | |
| Unconjugated | |
| TIMELESS | |
| C77407; Debt69; hTIM; mTim; Protein timeless homolog; RGD:620939}; rTIM; rTLP; TIM; Tim1; timeless; timeless {ECO:0000312; timeless circadian clock; timeless circadian clock 1; timeless circadian regulator; timeless homolog; TIMELESS1; timeless-like protein; Tof1 homolog; unknown | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 21853, 83508, 8914 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q9R1X4, Q9UNS1, Q9Z2Y1 | |
| TIMELESS | |
| A synthesized peptide derived from human TIMELESS(Accession Q9UNS1), corresponding to amino acid residues L1091-H1141. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction