missing translation for 'onlineSavingsMsg'
Learn More

Tim23 Rabbit Recombinant Antibody, Proteintech

Rabbit Recombinant Antibody

Brand:  Proteintech 82540-1-RR-100UL

 View more versions of this product

Product Code. 30237628

  • ONLINE EXCLUSIVE PRICE
    £360.00
    £330.00 / 100µL
    Save £30.00
    8% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Tim23 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Tim23. This antibody recognizes human, mouse, rat antigen. The Tim23 antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.
Specifications

Specifications

Tim23
Recombinant Monoclonal
500 μg/mL
MGC22767; PRO1197; TIM23
Recombinant Protein
100 μL
Primary
Human, Mouse, Rat
22 kDa
Liquid
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA
1H16
Unconjugated
Rabbit
Protein A purification
RUO
10431
Store at -20°C. Stable for one year after shipment.
Recombinant Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.