missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TIM-4 Monoclonal Antibody (RMT4-54)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA528256

Product Code. 16302534

  • £278.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody recognizes mouse Tim-4 (T-cell immunoglobulin mucin 4), a transmembrane glycoprotein and member of the immunoglobulin superfamily, exclusively expressed by antigen-presenting cells, which acts as the natural ligand of Tim-1, involved in T-cell expansion and cytokine production.

The T cell immunoglobulin and mucin domain containing protein (TIM) family encodes cell surface receptors that are involved in the regulation of T helper (Th) -1 and -2 cell-mediated immunity. TIM-4, which is preferentially expressed on macrophages and dendritic cells, is the natural ligand of TIM-1, and this binding leads to T-cell expansion and cytokine production. Unlike other members of the TIM family, TIM-4 lacks a putative tyrosine phosphorylation signal sequence in its intracellular domain. The TIM-4 gene maps to a locus associated with predisposition to asthma in both mice and humans and with its connection to TIM-1-triggered Th2 responsiveness, may be considered as a candidate disease/predisposition gene for asthma.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIM-4
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Q6U7R4
TIMD4
Tim-4-Ig, consisting of amino acids 1-288 of mouse Tim-4, coupled with the Fc portion of mouse IgG2a.
100 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
RMT4-54
Unconjugated
TIMD4
B430010N18Rik; RGD1564516; smuckler; soluble TIM 4; Spleen, mucin-containing, knockout of lymphotoxin protein; sTIM 4; T cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain containing 4; T-cell immunoglobulin and mucin domain-containing protein 4; T-cell immunoglobulin mucin receptor 4; T-cell membrane protein 4; Tim4; TIM-4; TIMD4; TIMD-4
Rat
Protein G
RUO
276891
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.