missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TIM-4 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
£147.94 - £422.75
Specifications
Antigen | TIM-4 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18739067
|
R&D Systems
AF2826-SP |
25 μg |
£147.94
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18089821
|
R&D Systems
AF2826 |
100 μg |
£422.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-4 Polyclonal specifically detects TIM-4 in Mouse samples. It is validated for Western Blot.Specifications
TIM-4 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD4, TIMD-4 | |
TIMD4 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q6U7R4 | |
91937 | |
Mouse myeloma cell line NS0-derived recombinant mouse TIM-4 Ala22-Thr279 Accession # Q6U7R4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse TIM-4 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) TIM-1, recombinant mouse (rm) TIM-2, rmTIM-3, rhTIM-4, rmTIM-5, and rmTIM-6 is observed. |
For Research Use Only