missing translation for 'onlineSavingsMsg'
Learn More

TIM-4 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18430606 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430606 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430606 Supplier Novus Biologicals Supplier No. NBP176702AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TIM-4 Polyclonal antibody specifically detects TIM-4 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-4
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the center of human TIM-4. The immunogen is located within amino acids 60 - 110 of TIM-4. Amino Acid Squence: RTDGMRVTSRKSAKYR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 91937
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.