missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-4 Antibody (921832), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2929F
436.05 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
TIM-4 Monoclonal antibody specifically detects TIM-4 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TIM-4 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 921832 | |
| Flow Cytometry, CyTOF-ready | |
| FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 | |
| Mouse myeloma cell line NSO-derived recombinant human TIM-4, Glu25-Leu315, Accession # Q96H15 | |
| 0.1 mL | |
| Cellular Signaling, Immunology | |
| 91937 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering