missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | TIM-4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 5G3 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry |
| Formulation | PBS |
| Gene Alias | FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4 |
| Gene Symbols | TIMD4 |
| Host Species | Rat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?