missing translation for 'onlineSavingsMsg'
Learn More

TIM-4 Antibody (5G3), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18343551 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343551 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343551 Supplier Novus Biologicals Supplier No. NBP281121AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TIM-4 Monoclonal specifically detects TIM-4 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-4
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 5G3
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ27515, SMUCKLER, T-cell immunoglobulin and mucin domain containing 4, T-cell immunoglobulin and mucin domain-containing protein 4, T-cell membrane protein 4, TIM4, TIM-4, TIMD-4
Gene Symbols TIMD4
Host Species Rabbit
Immunogen This antibody was raised by immunising female Lewis rats (Harlan Sprague-Dawley) with Tim-4:Ig fusion protein in complete Freund's adjuvant.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91937
Test Specificity This antibody is specific for murine TIM-4, which unlike other TIM family members, is not expressed on T lymphocytes but rather found on antigen presenting cells such as lymphoid and myeloid DC's and macrophages. TIM-4 has been shown to interact with TIM-1 thereby suggesting a role of TIM-4 in the balance of Th1/Th2 regulation. Additionally TIM-4 has been shown to be a receptor for phosphatidylserine and may play a role in engulfment of apoptotic cells.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.