missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 Rabbit anti-Human, Clone: 108, Novus Biologicals™
Rabbit Monoclonal Antibody
£287.00 - £420.00
Specifications
| Antigen | TIM-3 |
|---|---|
| Clone | 108 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18215057
|
Novus Biologicals
NBP2-89463-50UL |
50 μg |
£287.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18243336
|
Novus Biologicals
NBP2-89463-100UL |
100 μg |
£420.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-3 Monoclonal specifically detects TIM-3 in Human samples. It is validated for ELISA.Specifications
| TIM-3 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3 | |
| Recombinant Human TIM-3 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 108 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 84868 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title