missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIM-3 (CD366) Mouse anti-Human, Alexa Fluor 647, Clone: 7D3, BD Pharmingen™
Mouse Monoclonal Antibody
£113.93 - £267.41
Specifications
| Antigen | TIM-3 (CD366) |
|---|---|
| Clone | 7D3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15894389
|
BD Biosciences
565559 |
25 Tests |
£119.00 £113.93 / Each Save £5.07 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15884389
|
BD Biosciences
565558 |
100 Tests |
£279.00 £267.41 / Each Save £11.59 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.
TIM-3 (CD366) Monoclonal antibody specifically detects TIM-3 (CD366) in Human samples. It is validated for Flow cytometry.Specifications
| TIM-3 (CD366) | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human TIM-3 | |
| Primary | |
| Vol. Per test: 5μL |
| 7D3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD366; HAVCR2; TIM3; T cell immunoglobulin mucin-3; TIMD-3; KIM-3 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title