missing translation for 'onlineSavingsMsg'
Learn More

TIM-3 Antibody (344801) [DyLight 755], Novus Biologicals™

Product Code. 30737861 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737861 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737861 Supplier Novus Biologicals Supplier No. FAB23651Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

TIM-3 Monoclonal antibody specifically detects TIM-3 in Human samples. It is validated for Agonist Activity
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-3
Applications Agonist Activity
Classification Monoclonal
Clone 344801
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias CD366, FLJ14428, HAVcr-2, hepatitis A virus cellular receptor 2, kidney injury molecule-3, T cell immunoglobulin mucin 3, T cell immunoglobulin mucin-3, T-cell immunoglobulin and mucin domain-containing protein 3, TIM 3, Tim-3, TIM3 T-cell membrane protein 3, TIMD-3, TIMD3KIM-3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant human TIM-3, Met1-Arg200, Accession # Q8TDQ0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 84868
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.