missing translation for 'onlineSavingsMsg'
Learn More

TIM-1/KIM-1/HAVCR Antibody (3B3), DyLight 594, Novus Biologicals™

Product Code. 18320160 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18320160 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18320160 Supplier Novus Biologicals Supplier No. NBP281119DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIM-1/KIM-1/HAVCR
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 3B3
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Gene Symbols HAVCR1
Host Species Rabbit
Immunogen This antibody was raised by immunising female Lewis strain rats (Harlan Sprague-Dawley) subcutaneously with mouse TIM-1/KIM-1/HAVCR-Ig in complete Freund's adjuvant (CFA), followed by multiple 'boosting' with mouse TIM-1/KIM-1/HAVCR-Ig in PBS.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26762
Test Specificity This antibody is specific for murine TIM-1/KIM-1/HAVCR, a member of the T cell immunoglobulin domain, mucin-like domain (TIM) gene family. TIM-1/KIM-1/HAVCR has been found on activated CD4+ T cells, mast cells, and a subset of B cells. TIM-1/KIM-1/HAVCR is involved in renal injury, tissue homeostasis, and T cell costimulation, and promotes the differentiation of Th2 cells and the production of IL-4. TIM-1/KIM-1/HAVCR can bind several ligands, including Hepatitis A virus, phosphatidylserine, TIM4, IgA, as well as itself.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.