missing translation for 'onlineSavingsMsg'
Learn More

Tilfrinib, MedChemExpress

Product Code. 16447087
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447087 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16447087 Supplier Medchem Express Supplier No. HY110244
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tilfrinib (compound 4f) is a potent and selective inhibitor of breast tumor kinase (Brk) with an IC50 of 3.15 nM, which displays anti-proliferative activity and acts as a promising antitumor agent.

Tilfrinib (compound 4f) is a potent and selective inhibitor of breast tumor kinase (Brk) with an IC50 of 3.15 nM, which displays anti-proliferative activity and acts as a promising antitumor agent.

Specifications

Chemical Name or Material Tilfrinib
CAS 1600515-49-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H13N3O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (454.05 mM; Need ultrasonic)
SMILES OC1=CC=CC(NC2=C3C4=CC=CC=C4NC3=NC=C2)=C1
Molecular Weight (g/mol) 275.3
Formula Weight 275.3
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.