missing translation for 'onlineSavingsMsg'
Learn More

Tigogenin, TRC

Product Code. 30457167
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457167 250 mg 250mg
30450721 100 mg 100mg
30454260 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457167 Supplier Toronto Research Chemicals Supplier No. TRCT444685250MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-60-1
Molecular Formula C27H44O3
Molecular Weight (g/mol) 416.64
Synonym 5α-Spirostan-3β-ol, (25R)-5α-Spirostan-3β-ol, Tigogenin, (25R)-5α-Spirostan-3β-ol, NSC 93754, Ticogenin, (3β,5α,25R)-Spirostan-3-ol
SMILES C[C@@H]1CC[C@@]2(OC1)O[C@H]3C[C@H]4[C@@H]5CC[C@H]6C[C@@H](O)CC[C@]6(C)[C@H]5CC[C@]4(C)[C@H]3[C@@H]2C

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C27H44O3/c1-16-7-12-27(29-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(28)8-10-25(18,3)21(20)9-11-26(22,24)4/h16-24,28H,5-15H2,1-4H3/t16-,17+,18+,19+,20-,21+,22+,23+,24+,25+,26+,27-/m1/s1
Formula Weight 416.329
Chemical Name or Material Tigogenin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.