missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Mouse, PE, Clone: 1G9, enQuireBio™
Mouse Monoclonal Antibody
£109.00 - £256.00
Specifications
Antigen | TIGIT |
---|---|
Clone | 1G9 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952228
|
EnquireBio
QAB57-PE-25UG |
25 μg |
£109.00
25µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942228
|
EnquireBio
QAB57-PE-100UG |
100 μg |
£256.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TIGIT | |
0.2mg/mL | |
Monoclonal | |
Mouse | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
100043314 | |
IgG1 κ | |
Protein A/G Purified |
1G9 | |
Flow Cytometry | |
PE | |
RUO | |
XP_017172323 | |
Tigit | |
Primary | |
Store at 2-4°C. |