missing translation for 'onlineSavingsMsg'
Learn More

TIGIT, Mouse anti-Human, Clone: TIGIT/3106, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79794-100UG

 View more versions of this product

Product Code. 18285607

  • £403.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TIGIT Monoclonal specifically detects TIGIT in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TIGIT
Monoclonal
0.2 mg/mL
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Mouse
Protein A or G purified
RUO
201633
Store at 4°C.
IgG2b κ
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
TIGIT/3106
Unconjugated
10 mM PBS with 0.05% BSA
TIGIT
Recombinant fragment (around aa 22-141) of human TIGIT protein (exact sequence is proprietary)
100 μg
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.