Learn More
TIGIT Monoclonal Antibody (GIGD7), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 25-9501-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The GIGD7 monoclonal antibody reacts with mouse T cell immunoreceptor with Ig and ITIM domains (TIGIT, VSTM3, WUCAM), a 26 kDa protein that is a member of the poliovirus receptor (PVR) family. In mice, the expression of TIGIT has been reported in follicular T helper cells, while in humans, the expression of TIGIT has been reported on NK cells, regulatory T cells, follicular T helper cells, memory CD4+ T cells, and CD8+ T cells. TIGIT is not expressed on B cells or naive CD4+ T cells. TIGIT is upregulated on CD4+ T cells following activation. TIGIT can interact with certain members of the PVR and PVR-like families, including CD155, and also mediates the interaction of NK cells and T cells with antigen presenting cells, fibroblasts and endothelial cells that express PVR and PVR-like proteins. Applications Reported: This GIGD7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This GIGD7 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation.
TIGIT Monoclonal antibody specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry.Specifications
TIGIT | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P86176 | |
TIGIT | |
mTIGIT-Fc treated with 4% PFA | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
Flow Cytometry | |
GIGD7 | |
PE-Cyanine7 | |
TIGIT | |
ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
Rat | |
Affinity chromatography | |
RUO | |
100043314 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
For Research Use Only.