missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGIT Monoclonal Antibody (GIGD7), Brilliant Violet™ 421, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£160.00 - £327.00
Specifications
| Antigen | TIGIT |
|---|---|
| Clone | GIGD7 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17922161
|
Invitrogen
404-9501-82 |
100 μg |
£327.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17902051
|
Invitrogen
404-9501-80 |
25 μg |
£160.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.Specifications
| TIGIT | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| ENSMUSG00000071552; T cell immunoreceptor with Ig and ITIM domains; T-cell immunoreceptor with Ig and ITIM domains; TIGIT; V-set and immunoglobulin domain containing 9; V-set and immunoglobulin domain-containing protein 9; V-set and transmembrane domain containing 3; V-set and transmembrane domain-containing protein 3; VSIG9; VSTM3; Washington University cell adhesion molecule; WUCAM | |
| TIGIT | |
| IgG2a κ | |
| Affinity chromatography | |
| TIGIT |
| GIGD7 | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Rat | |
| Mouse | |
| P86176 | |
| 100043314 | |
| mTIGIT-Fc treated with 4% PFA | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title