missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Antibody (4D4), DyLight 680, Novus Biologicals™

Product Code. 18373390 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373390 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373390 Supplier Novus Biologicals Supplier No. NBP281116FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Armenian Hamster Monoclonal Antibody

TIGIT Monoclonal specifically detects TIGIT in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TIGIT
Applications Flow Cytometry
Classification Monoclonal
Clone 4D4
Conjugate DyLight 680
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias T cell immunoreceptor with Ig and ITIM domains, TIGIT, VSIG9, VSTM3, WUCAM
Gene Symbols TIGIT
Host Species Armenian Hamster
Immunogen Armenian hamsters were immunized with recombinant mouse TIGIT tetramers by a combination of s.c. and food pad immunization and booster injections. Draining lymph nodes were fused with Sp2/0-Ag14 cells, selected in HAT (hypoxanthine/aminopterin/thymidine) medium and supernatants were screened for specificity by ELISA and flow cytometry using TIGIT-transfectants.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201633
Test Specificity 4D4 specifically recognises TIGIT.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.