missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559532
This item is not returnable.
View return policy
Description
Immunogen sequence: MNKRGKYTTL NLEEKMKVLS RIEAGRSLKS VMDEFGISKS TFYDIKKNKK LILDFVLKQD MPLVGAEKRK RTTGAKYGDV DDAVYMWYQQ KRSAG Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 34%.
The protein encoded by this gene belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.
Specifications
| TIGD7 | |
| Polyclonal | |
| Unconjugated | |
| TIGD7 | |
| Sancho; TIGD7; tigger transposable element derived 7; tigger transposable element-derived protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91151 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NT04 | |
| TIGD7 | |
| Recombinant protein corresponding to Human TIGD7. Recombinant protein control fragment (Product #RP-98735). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction