missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TIGD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567151
This item is not returnable.
View return policy
Description
Immunogen sequence: KSSTITKAWKK LFPGNEENSG MNIDEGAILA ANLATVLQNT EECEHVDIEN IDQWFDSRSS DSSCQVLTD Highest antigen sequence identity to the following orthologs - mouse 81%, rat 81%.
TIGD2 belongs to the tigger subfamily of the pogo superfamily of DNA-mediated transposons in humans. These proteins are related to DNA transposons found in fungi and nematodes, and more distantly to the Tc1 and mariner transposases. They are also very similar to the major mammalian centromere protein B. The exact function of this gene is not known.
Specifications
| TIGD2 | |
| Polyclonal | |
| Unconjugated | |
| TIGD2 | |
| 3632410O17Rik; epididymis luminal protein 106; HEL106; LOC100912924; Tigd2; tigger transposable element derived 2; tigger transposable element derived 2 homolog; tigger transposable element-derived protein 2; uncharacterized LOC100912924 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 166815 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4W5G0 | |
| TIGD2 | |
| Recombinant Human TIGD2. Recombinant protein control fragment (Product #RP-105522). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction